Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172563.3 | NP_766151.1 | 1, 2 | 665 | ||
GenBank mRNA | AK054062.1 | - | 1, 2 | 155 | ||
BC058705.1 | - | 1, 2 | 643 | 6839029 |