Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291765.1 | NP_001278694.1 | 7 | 627 | ||
NM_001291766.1 | NP_001278695.1 | 6 | 621 | |||
NM_001291768.1 | NP_001278697.1 | 6 | 560 | |||
NM_009361.3 | NP_033387.1 | 7 | 688 | |||
GenBank mRNA | AK013180.1 | - | 6 | 590 | ||
AK019997.1 | - | 7 | 628 | |||
AK030820.1 | - | 7 | 689 | |||
AK154037.1 | - | 6 | 528 | |||
AK198738.1 | - | 1 | 13 | |||
AK207929.1 | - | 1 | 13 | |||
BC058633.1 | - | 6 | 903 | |||
BC132567.1 | - | 6 | 533 | 40130844 | ||
BC132569.1 | - | 6 | 533 | 40130846 | ||
BC145344.1 | - | 6 | 533 | 9053900 | ||
X72310.1 | - | 6 | 562 |