Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011576.1 | NP_035706.1 | 8 | 1145 | ||
GenBank mRNA | AK028356.1 | - | 8 | 1145 | ||
AK164207.1 | - | 1 | 3849 | |||
AK167522.1 | - | 8 | 1007 |