Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145460.2 | NP_663435.2 | 6, 7 | 1000 | ||
GenBank mRNA | AK031978.1 | - | 5, 6 | 629 | ||
AK075924.1 | - | 5, 6 | 626 | |||
AK139884.1 | - | 6, 7 | 1000 | |||
BC019806.1 | - | 6, 7 | 778 | 4018387 |