Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009415.2 | NP_033441.2 | 1 | 249 | ||
GenBank mRNA | AK002270.1 | - | 1 | 112 | ||
AK010808.1 | - | 1 | 119 | |||
AK053780.1 | - | 1 | 108 | |||
AK146013.1 | - | 1 | 108 | |||
AK149768.1 | - | 1 | 113 | |||
AK150735.1 | - | 1 | 110 | |||
AK159741.1 | - | 1 | 113 | |||
AK167437.1 | - | 1 | 114 | |||
AK168060.1 | - | 1 | 112 | |||
AK168446.1 | - | 1 | 113 | |||
AK168756.1 | - | 1 | 112 | |||
BC004024.1 | - | 1 | 97 | |||
BC046761.1 | - | 1 | 100 | 5693696 | ||
BU945752.1 | - | 1 | 254 | |||
L31793.1 | - | 1 | 106 | |||
X53333.1 | - | 1 | 111 |