Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011655.5 | NP_035785.1 | 4 | 815 | ||
GenBank mRNA | AK011263.1 | - | 4 | 743 | ||
AK011765.1 | - | 4 | 747 | |||
AK012350.1 | - | 4 | 747 | |||
AK012365.1 | - | 2 | 385 | |||
AK013283.1 | - | 2 | 375 | |||
AK017806.1 | - | 4 | 745 | |||
AK051164.1 | - | 4 | 747 | |||
AK051393.1 | - | 4 | 749 | |||
AK083327.1 | - | 4 | 747 | |||
AK146567.1 | - | 4 | 747 | |||
AK148978.1 | - | 4 | 742 | |||
AK151215.1 | - | 4 | 747 | |||
AK151670.1 | - | 4 | 747 | |||
AK160225.1 | - | 4 | 747 | |||
AK161575.1 | - | 4 | 747 | |||
AK165249.1 | - | 4 | 745 | |||
AK167691.1 | - | 4 | 747 | |||
AK167779.1 | - | 4 | 747 | |||
AK167808.1 | - | 4 | 742 | |||
AK167926.1 | - | 4 | 747 | |||
AK168271.1 | - | 4 | 742 | |||
AK169123.1 | - | 4 | 742 | |||
AK169210.1 | - | 4 | 746 | |||
AK169295.1 | - | 4 | 747 | |||
AK169398.1 | - | 1 | 224 | |||
AK169606.1 | - | 4 | 747 | |||
AK186299.1 | - | 1 | 102 | |||
AK188125.1 | - | 1 | 103 | |||
AK205152.1 | - | 1 | 102 | |||
BC003825.1 | - | 4 | 721 | 3601177 | ||
X04663.1 | - | 4 | 645 |