Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144845.3 | NP_659094.1 | 4 | 738 | ||
GenBank mRNA | AK041045.1 | - | 4 | 723 | ||
AK050128.1 | - | 4 | 739 | |||
AK052644.1 | - | 4 | 748 | |||
AK143815.1 | - | 4 | 753 | |||
BC022134.1 | - | 4 | 723 | 5098754 | ||
BC034837.1 | - | 4 | 719 | 5134131 |