Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009504.4 | NP_033530.2 | 7 | 818 | ||
GenBank mRNA | AK030339.1 | - | 7 | 819 | ||
AK154647.1 | - | 7 | 793 | |||
AK156789.1 | - | 7 | 834 | |||
BC006716.1 | - | 7 | 784 | 3710866 | ||
CT010294.1 | - | 5 | 678 | |||
D31969.1 | - | 7 | 786 |