Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011701.4 | NP_035831.2 | 1 | 402 | ||
GenBank mRNA | AK033175.1 | - | 1 | 190 | ||
AK076001.1 | - | 1 | 404 | |||
AK131618.1 | - | 1 | 404 | |||
AK133726.1 | - | 1 | 403 | |||
AK150270.1 | - | 1 | 98 | |||
AK151196.1 | - | 1 | 405 | |||
AK153019.1 | - | 1 | 405 | |||
AK159539.1 | - | 1 | 403 | |||
AK169181.1 | - | 1 | 404 | |||
AK170429.1 | - | 1 | 403 | |||
AK205778.1 | - | 1 | 50 | |||
AK208041.1 | - | 1 | 192 | |||
AK217953.1 | - | 1 | 248 | |||
BC089335.1 | - | 1 | 398 | 6415629 | ||
BC134386.1 | - | 1 | 303 | |||
M24849.1 | - | 1 | 319 | |||
M26251.1 | - | 1 | 761 | |||
X51438.1 | - | 1 | 371 | |||
X56397.1 | - | 1 | 365 |