Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011701.4 | NP_035831.2 | 8, 9 | 1490 | ||
GenBank mRNA | AK012469.1 | - | 8, 9 | 993 | ||
AK076001.1 | - | 8, 9 | 1487 | |||
AK131618.1 | - | 8, 9 | 1492 | |||
AK133726.1 | - | 8, 9 | 1491 | |||
AK150270.1 | - | 8, 9 | 1186 | |||
AK151196.1 | - | 8, 9 | 1493 | |||
AK153019.1 | - | 8, 9 | 1493 | |||
AK169181.1 | - | 8, 9 | 1492 | |||
AK170429.1 | - | 8, 9 | 1491 | |||
AK203121.1 | - | 3, 4 | 159 | |||
AK205443.1 | - | 3, 4 | 281 | |||
BC089335.1 | - | 8, 9 | 1486 | 6415629 | ||
M24849.1 | - | 8, 9 | 1407 | |||
M26251.1 | - | 8, 9 | 1849 | |||
X51438.1 | - | 8, 9 | 1459 | |||
X56397.1 | - | 8, 9 | 1453 |