Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029843.1 | NP_001025014.1 | 9 | 823 | ||
NM_001029844.1 | NP_001025015.1 | 9 | 823 | |||
NM_011705.3 | NP_035835.1 | 9 | 823 | |||
GenBank mRNA | AF080252.1 | - | 8 | 736 | ||
AF080253.1 | - | 9 | 777 | |||
AK032344.1 | - | 9 | 794 | |||
AK039554.1 | - | 3 | 195 | |||
AK088230.1 | - | 9 | 747 | |||
AK088674.1 | - | 9 | 806 | |||
AK136352.1 | - | 9 | 797 | |||
AK166676.1 | - | 9 | 791 | |||
BC016676.1 | - | 9 | 858 | 3602292 | ||
BC051072.1 | - | 9 | 803 |