Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011711.1 | NP_035841.1 | 18 | 2188 | ||
GenBank mRNA | AK129473.1 | - | 18 | 2107 | ||
AK139711.1 | - | 18 | 2172 | |||
AK143308.1 | - | 18 | 2088 | |||
AK155331.1 | - | 18 | 2205 | |||
AK163361.1 | - | 18 | 2175 | |||
AK170744.1 | - | 17 | 2018 | |||
BC023797.1 | - | 17 | 1908 | |||
BC027845.1 | - | 2 | 165 | |||
BC085094.1 | - | 18 | 2083 | |||
BC131961.1 | - | 18 | 2064 | 40130238 |