Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163307.1 | NP_001156779.1 | 7 | 677 | ||
NM_001163308.1 | NP_001156780.1 | 6 | 539 | |||
NM_009545.2 | NP_033571.1 | 9 | 944 | |||
GenBank mRNA | AF483502.1 | - | 5 | 385 | ||
AF483503.1 | - | 5 | 385 | |||
AK142837.1 | - | 6 | 452 | |||
BC016419.1 | - | 7 | 631 | 4458285 | ||
D90085.1 | - | 9 | 792 |