Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011756.4 | NP_035886.1 | 2 | 117 | ||
GenBank mRNA | AK005006.1 | - | 1 | 825 | ||
AK088525.1 | - | 1 | 825 | |||
AK154825.1 | - | 2 | 145 | |||
BC021391.1 | - | 2 | 116 | 5053664 | ||
M57422.1 | - | 2 | 117 | |||
M58565.1 | - | 2 | 86 | |||
M58691.1 | - | 1141 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK158286.1 | - | 1860 | Chr. - - [-] on Build |