Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm4302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166634.1 | NP_001160106.1 | 1 | 67 | ||
GenBank mRNA | BC158099.1 | - | 1 | 67 | 9007384 |
Gene Symbol: Gm4303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166638.1 | NP_001160110.1 | 1 | 65 | ||
GenBank mRNA | BC158090.1 | - | 1 | 65 | 9007379 |
Gene Symbol: Gm4305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166639.1 | NP_001160111.1 | 65 |
Gene Symbol: Gm4307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166641.1 | NP_001160113.1 | 65 |
Gene Symbol: Gm4850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG069969.2 | - | 1 | 695 |