Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177627.1 | NP_001171098.1 | 2 | 276 | ||
NM_172659.2 | NP_766247.2 | 2 | 276 | |||
GenBank mRNA | AK079650.1 | - | 2 | 220 | ||
AK089246.1 | - | 2 | 268 | |||
AK137441.1 | - | 2 | 242 | |||
AK150104.1 | - | 2 | 229 | |||
AK163445.1 | - | 2 | 238 | |||
AK166312.1 | - | 2 | 234 | |||
AK167904.1 | - | 2 | 238 | |||
AK172411.1 | - | 2 | 220 | |||
BC058210.1 | - | 2 | 200 | |||
BC141168.1 | - | 2 | 207 | 9055780 | ||
BC145480.1 | - | 2 | 207 | |||
BY202464.1 | - | 2 | 277 |