Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036833.1 | - | 8 | 1338 | ||
AK077815.1 | - | 8 | 1666 | |||
AK142856.1 | - | 6 | 865 | |||
AK220235.1 | - | 8 | 1334 | |||
AY536269.1 | - | 4 | 543 | |||
BC138789.1 | - | 7 | 899 | 8861811 | ||
L03547.1 | - | 7 | 884 |