Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045807.1 | NP_001039272.1 | 1 | 338 | ||
GenBank mRNA | AK048987.1 | - | 89 | |||
AK077048.1 | - | 1 | 723 | |||
AK166271.1 | - | 1 | 101 | |||
AK168242.1 | - | 1 | 99 | |||
AK168936.1 | - | 1 | 223 | |||
BC051409.1 | - | 1 | 92 | |||
BC057038.1 | - | 1 | 102 | |||
BC080828.1 | - | 1 | 83 | |||
BC120590.1 | - | 1 | 338 | 40129513 | ||
BC137741.1 | - | 1 | 338 | 8860760 |