Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045807.1 | NP_001039272.1 | 1 | 856 | ||
GenBank mRNA | AK077048.1 | - | 1 | 1241 | ||
AK166271.1 | - | 1 | 619 | |||
AK168242.1 | - | 1 | 617 | |||
AK168936.1 | - | 1 | 741 | |||
AK220162.1 | - | 1 | 668 | |||
BC051409.1 | - | 1 | 610 | |||
BC057038.1 | - | 1 | 620 | |||
BC080828.1 | - | 1 | 601 | |||
BC120590.1 | - | 1 | 856 | 40129513 | ||
BC137741.1 | - | 1 | 856 | 8860760 |