Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029867.1 | NP_001025038.1 | 1 | 134 | ||
GenBank mRNA | AK149425.1 | - | 1 | 137 | ||
BC141232.1 | - | 1 | 129 | 9055844 | ||
BC145555.1 | - | 1 | 129 | 9054119 |