Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130309D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172726.4 | NP_766314.3 | 4 | 1111 | ||
GenBank mRNA | AK041667.1 | - | 4 | 1057 | ||
AK050733.1 | - | 4 | 1025 | |||
AK053801.1 | - | 4 | 1112 | |||
AK054218.1 | - | 4 | 812 | |||
AK087531.1 | - | 4 | 1112 | |||
AK172614.1 | - | 4 | 1058 | |||
AK178927.1 | - | 1 | 155 | |||
AK198635.1 | - | 2 | 298 | |||
AK212039.1 | - | 1 | 105 | |||
BC058940.1 | - | 3 | 3190 | |||
BC079853.1 | - | 4 | 1045 | 5716912 | ||
BC082321.1 | - | 4 | 855 | 6411746 |