Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130309D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172726.4 | NP_766314.3 | 4, 5 | 1373 | ||
GenBank mRNA | AK041667.1 | - | 4, 5 | 1319 | ||
AK050733.1 | - | 4, 5 | 1287 | |||
AK053801.1 | - | 4, 5 | 1374 | |||
AK054218.1 | - | 4, 5 | 1074 | |||
AK087531.1 | - | 4, 5 | 1374 | |||
AK172614.1 | - | 4, 5 | 1320 | |||
AK185519.1 | - | 1, 2 | 29 | |||
BC058940.1 | - | 3, 4 | 3452 | |||
BC079853.1 | - | 4, 5 | 1307 | 5716912 | ||
BC082321.1 | - | 4, 5 | 1117 | 6411746 |