Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153572.2 | NP_705800.1 | 2 | 158 | ||
GenBank mRNA | AK049620.1 | - | 2 | 512 | ||
BC030434.1 | - | 2 | 145 | 5369313 | ||
BE653455.1 | - | 2 | 158 |