Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139925.1 | - | 3 | 885 | ||
AK145355.1 | - | 2 | 574 | |||
AY364009.1 | - | 800 | ||||
AY596202.1 | - | 2 | 800 |