Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139925.1 | - | 3 | 1333 | ||
AK145355.1 | - | 2 | 1022 | |||
AY364009.1 | - | 1248 | ||||
AY596202.1 | - | 2 | 1248 |