Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183312.3 | NP_899135.3 | 2 | 936 | ||
NM_201639.2 | NP_964001.2 | 2 | 936 | |||
NM_207663.3 | NP_997546.2 | 2 | 936 | |||
GenBank mRNA | AJ579700.1 | - | 2 | 887 | ||
AJ579701.1 | - | 2 | 887 | |||
AJ579702.1 | - | 2 | 887 | |||
AK143019.1 | - | 2 | 937 | |||
AK172937.1 | - | 1 | 17 | |||
BC026872.1 | - | 3 | 780 | |||
BC040093.1 | - | 3 | 780 |