Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163713.1 | NP_001157185.1 | 3 | 417 | ||
NM_172745.3 | NP_766333.1 | 3 | 417 | |||
GenBank mRNA | AK075857.1 | - | 3 | 341 | ||
AK084724.1 | - | 3 | 398 | |||
AK152858.1 | - | 3 | 412 | |||
AK153135.1 | - | 3 | 412 | |||
BC060959.1 | - | 3 | 394 | 30379757 | ||
BC100596.1 | - | 3 | 410 | 6762419 | ||
BY246546.1 | - | 3 | 417 |