Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027626.1 | NP_081902.1 | 6 | 830 | ||
NM_030263.5 | NP_084539.2 | 11 | 1172 | |||
NM_177698.4 | NP_808366.2 | 14 | 2936 | |||
GenBank mRNA | AB220685.1 | - | 14 | 2936 | ||
AK031477.1 | - | 6 | 773 | |||
AK122405.1 | - | 5 | 486 | |||
AK147606.1 | - | 6 | 830 | |||
AK161498.1 | - | 11 | 1158 | |||
BC042208.1 | - | 2 | 159 | |||
BC052365.1 | - | 11 | 1137 | |||
BC131913.1 | - | 6 | 742 | |||
BC145641.1 | - | 6 | 742 |