Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144929.2 | NP_659178.1 | 4, 5 | 1223 | ||
GenBank mRNA | AK039726.1 | - | 2, 3 | 1730 | ||
AK080715.1 | - | 4, 5 | 539 | |||
AY428631.1 | - | 3, 4 | 435 | |||
AY527275.1 | - | 4, 5 | 1223 | |||
AY527276.1 | - | 5, 6 | 678 | |||
BC022744.1 | - | 3, 4 | 795 | 4527012 | ||
BC023051.1 | - | 3, 4 | 751 | 5361063 | ||
BC027079.1 | - | 3, 4 | 751 | 5357069 |