Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144930.2 | NP_659179.1 | 10 | 1129 | ||
GenBank mRNA | AB023632.1 | - | 6 | 532 | ||
AK136491.1 | - | 10 | 1121 | |||
BC013479.1 | - | 10 | 1104 | 4239756 | ||
EH107325.1 | - | 5 | 377 |