Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144930.2 | NP_659179.1 | 5 | 713 | ||
GenBank mRNA | AB023632.1 | - | 1 | 116 | ||
AI786051.1 | - | 1 | 109 | |||
AK136491.1 | - | 5 | 705 | |||
BC013479.1 | - | 5 | 688 | 4239756 |