Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144936.1 | NP_659185.1 | 4, 5 | 671 | ||
GenBank mRNA | AK079262.1 | - | 4, 5 | 658 | ||
AK144325.1 | - | 4, 5 | 896 | |||
BC018222.1 | - | 4, 5 | 627 | 4221959 | ||
BC026654.1 | - | 4, 5 | 671 | 4988382 |