Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fez1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183171.4 | NP_898994.2 | 4, 5 | 714 | ||
GenBank mRNA | BC029629.1 | - | 5, 6 | 819 | 4035444 | |
DQ141674.1 | - | 4, 5 | 718 |