Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430571L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175486.3 | NP_780695.1 | 4, 5 | 660 | ||
GenBank mRNA | AK032358.1 | - | 4, 5 | 661 | ||
AK032501.1 | - | 4, 5 | 667 | |||
AK043338.1 | - | 4, 5 | 700 | |||
AK085251.1 | - | 4, 5 | 691 | |||
BC115775.1 | - | 4, 5 | 572 | 40107115 | ||
BC116708.1 | - | 4, 5 | 572 | 40107114 |