Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004145.2 | NP_001004145.1 | 2180 | |||
GenBank mRNA | AK136247.1 | - | 2544 | |||
BC080833.1 | - | 2180 | 30481413 |