Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177707.3 | NP_808375.1 | 12 | 1438 | ||
GenBank mRNA | AK036516.1 | - | 12 | 1439 | ||
BC067208.1 | - | 12 | 1218 | 30286602 |