Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011830.3 | NP_035960.2 | 7 | 835 | ||
GenBank mRNA | AK011777.1 | - | 7 | 863 | ||
AK143505.1 | - | 7 | 876 | |||
AK146362.1 | - | 7 | 901 | |||
AK150713.1 | - | 7 | 871 | |||
AK151233.1 | - | 7 | 871 | |||
AK151706.1 | - | 7 | 894 | |||
AK152691.1 | - | 7 | 868 | |||
AK159822.1 | - | 7 | 868 | |||
AK160569.1 | - | 7 | 868 | |||
AK166667.1 | - | 7 | 870 | |||
AK169129.1 | - | 7 | 901 | |||
AK169788.1 | - | 7 | 901 | |||
BC010314.1 | - | 7 | 835 | 2646679 | ||
BC052671.1 | - | 7 | 856 | 30058399 | ||
M33934.1 | - | 7 | 868 | |||
M98333.1 | - | 7 | 802 |