Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166251.1 | NP_001159723.1 | 6, 7 | 1062 | ||
NM_011844.4 | NP_035974.1 | 6, 7 | 747 | |||
GenBank mRNA | AJ001118.2 | - | 6, 7 | 675 | ||
AJ316580.1 | - | 6, 7 | 938 | |||
AK028008.1 | - | 6, 7 | 691 | |||
AK131645.1 | - | 6, 7 | 727 | |||
AK148242.1 | - | 6, 7 | 690 | |||
AK188789.1 | - | 2, 3 | 111 | |||
AK190641.1 | - | 2, 3 | 110 | |||
AK191024.1 | - | 2, 3 | 110 | |||
AK196565.1 | - | 2, 3 | 110 | |||
AK213989.1 | - | 3, 4 | 240 | |||
BC057965.1 | - | 6, 7 | 672 | 4986023 |