Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081409.1 | NP_001074878.1 | 9, 10 | 1244 | ||
GenBank mRNA | AK032638.1 | - | 9, 10 | 1259 | ||
BC052212.1 | - | 8, 9 | 1170 | |||
BC061492.1 | - | 8, 9 | 1110 | |||
BC082777.1 | - | 9, 10 | 1239 | |||
BC145689.1 | - | 8, 9 | 975 | 40130992 |