Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173769.3 | NP_776130.1 | 5 | 483 | ||
GenBank mRNA | AK036826.1 | - | 5 | 484 | ||
AK087530.1 | - | 5 | 531 | |||
AK136402.1 | - | 5 | 561 | |||
BC111896.1 | - | 5 | 441 | 40061302 | ||
BC112418.1 | - | 5 | 441 | 40061301 |