Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacsin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286743.1 | NP_001273672.1 | 9 | 1369 | ||
NM_001286744.1 | NP_001273673.1 | 9 | 1275 | |||
NM_011861.3 | NP_035991.1 | 9 | 1334 | |||
NM_178365.4 | NP_848142.1 | 9 | 1352 | |||
GenBank mRNA | AK043967.1 | - | 9 | 1338 | ||
AK077958.1 | - | 4 | 456 | |||
AK158686.1 | - | 9 | 1372 | |||
BC014698.1 | - | 9 | 1223 | 4527708 | ||
X85124.1 | - | 9 | 1243 |