Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172822.3 | NP_766410.2 | 3 | 1883 | ||
GenBank mRNA | AK087516.1 | - | 3 | 1769 | ||
AK089183.1 | - | 3 | 1782 | |||
BC117748.2 | - | 2 | 1703 | 40090477 |