Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011879.2 | NP_036009.1 | 12 | 1132 | ||
GenBank mRNA | AJ006130.1 | - | 12 | 1015 | ||
AK029644.1 | - | 12 | 1086 | |||
AK050625.1 | - | 12 | 1090 | |||
AK082356.1 | - | 12 | 1064 | |||
AK087976.1 | - | 12 | 1078 | |||
BC006706.1 | - | 10 | 1490 | |||
BC014739.1 | - | 12 | 1063 | 4920184 | ||
BC026569.1 | - | 12 | 1063 | 4982612 | ||
BC029148.1 | - | 10 | 2026 | |||
BC029671.1 | - | 12 | 1075 | 5364953 | ||
BC029694.1 | - | 12 | 1072 | 5369477 | ||
BC033356.1 | - | 12 | 1059 | 5366230 |