Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF176903.1 | - | 3 | 1262 | ||
AK014578.1 | - | 3 | 1280 | |||
AK140405.1 | - | 3 | 1274 | |||
BC039139.1 | - | 4 | 1120 | 3672353 | ||
BC051977.1 | - | 2 | 1566 | |||
BC069914.1 | - | 1 | 783 | 5251876 |