Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044747.2 | NP_001038212.1 | 5 | 1895 | ||
NM_001163797.1 | NP_001157269.1 | 6 | 1985 | |||
NM_013844.2 | NP_038872.1 | 6 | 1991 | |||
GenBank mRNA | AB024005.1 | - | 6 | 1975 | ||
AK013868.1 | - | 6 | 1986 | |||
AK044831.1 | - | 5 | 1865 | |||
AK143158.1 | - | 1 | 391 | |||
AK146092.1 | - | 6 | 1952 | |||
AK153887.1 | - | 6 | 1927 | |||
BC018171.1 | - | 6 | 1974 | 3989444 |