Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005342.2 | NP_001005342.1 | 4, 5 | 561 | ||
GenBank mRNA | AB098743.2 | - | 4, 5 | 561 | ||
AK043577.1 | - | 1, 2 | 399 | |||
BC119263.1 | - | 3, 4 | 435 | 8734012 |