Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173426.2 | NP_775602.1 | 3 | 1536 | ||
GenBank mRNA | AK052596.1 | - | 3 | 1537 | ||
AK077232.1 | - | 1 | 1169 | |||
AK146011.1 | - | 2 | 1294 | |||
AK159054.1 | - | 3 | 1545 | |||
BC055683.1 | - | 3 | 1466 | 6414160 |