Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163732.1 | NP_001157204.1 | 10 | 1042 | ||
NM_172869.4 | NP_766457.1 | 10 | 1042 | |||
GenBank mRNA | AK034956.1 | - | 10 | 1017 | ||
AK048561.1 | - | 10 | 985 | |||
AK079349.1 | - | 10 | 1046 | |||
BC037727.1 | - | 10 | 1213 | |||
BC112419.1 | - | 10 | 950 | 40061426 | ||
BC113795.1 | - | 10 | 950 | 40061428 | ||
BC118964.2 | - | 10 | 952 | 40097059 |