Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136471.1 | NP_001129943.1 | 3, 4 | 260 | ||
NM_172297.1 | NP_758501.2 | 2, 3 | 277 | |||
GenBank mRNA | AK134892.1 | - | 3, 4 | 246 | ||
BC021915.1 | - | 2, 3 | 277 | 4527178 | ||
BY029379.1 | - | 3, 4 | 298 |