Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136471.1 | NP_001129943.1 | 10 | 1114 | ||
NM_172297.1 | NP_758501.2 | 9 | 1131 | |||
GenBank mRNA | AK011186.1 | - | 3 | 1938 | ||
AK134892.1 | - | 10 | 1100 | |||
BC021915.1 | - | 9 | 1131 | 4527178 |